Product Details

SNP ID
rs148941896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93425008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAGATGAGACTGGAGATTTGGAG[A/G]GTTTCAATGTACCTCCTCATTTAGC
Phenotype
MIM: 114131
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CALCR PubMed Links

Gene Details

Gene
CALCR
Gene Name
calcitonin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164737.1 3056 UTR 3 NP_001158209.1
NM_001164738.1 3056 UTR 3 NP_001158210.1
NM_001742.3 3056 UTR 3 NP_001733.1

View Full Product Details