Product Details

SNP ID
rs149628324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93426534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGTTGGAGGGGCGCCTCCCCCAA[C/T]GCTGGTTCCACTGAATTTTGAATTG
Phenotype
MIM: 114131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALCR PubMed Links

Gene Details

Gene
CALCR
Gene Name
calcitonin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164737.1 1530 Missense Mutation CAT,CGT H450R NP_001158209.1
NM_001164738.1 1530 Missense Mutation CAT,CGT H416R NP_001158210.1
NM_001742.3 1530 Missense Mutation CAT,CGT H416R NP_001733.1

View Full Product Details