Product Details

SNP ID
rs141415713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:132714051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGCTGAGAAACAATGATTTCAC[A/G]TCTTGAGTGAAACAGAGAAAATATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM71 PubMed Links

Gene Details

Gene
TMEM71
Gene Name
transmembrane protein 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145153.1 988 Silent Mutation TAC,TAT Y209Y NP_001138625.1
NM_144649.2 988 Silent Mutation TAC,TAT Y253Y NP_653250.2

View Full Product Details