Product Details

SNP ID
rs142974098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30996664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATTTGACAACAGTGAAAAAGGT[G/T]GATATTCTCTCTGGATTTGGGGTAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PURG PubMed Links

Gene Details

Gene
PURG
Gene Name
purine rich element binding protein G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015508.2 1831 Missense Mutation AAC,CAC N300H NP_001015508.1
NM_001323311.1 1831 Intron NP_001310240.1
NM_001323312.1 1831 Missense Mutation AAC,CAC N300H NP_001310241.1
NM_013357.2 1831 Intron NP_037489.1

View Full Product Details