Product Details

SNP ID
rs143277641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:101198608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCAGTTATCTACAGGGTACACCT[A/G]ACTGGCAGTTAGAGCCAGGTCATAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF706 PubMed Links

Gene Details

Gene
ZNF706
Gene Name
zinc finger protein 706
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042510.1 1175 UTR 3 NP_001035975.1
NM_001267708.1 1175 UTR 3 NP_001254637.1
NM_001267709.1 1175 UTR 3 NP_001254638.1
NM_016096.3 1175 UTR 3 NP_057180.1

View Full Product Details