Product Details

SNP ID
rs144109404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123645805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGCATGGTCAAAGAGCTTGTG[C/T]GGCAGCTGTCCCTGGGATGTCCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL38 PubMed Links

Gene Details

Gene
KLHL38
Gene Name
kelch like family member 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081675.2 1925 Silent Mutation CCA,CCG P560P NP_001075144.2
XM_005250901.4 1925 Silent Mutation CCA,CCG P560P XP_005250958.1

View Full Product Details