Product Details

SNP ID
rs146656524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:21909449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCAGCTGTCGCCTGCCTCCTCCA[C/T]GCCTCACCCCGGGGCTCCTGCGGGC
Phenotype
MIM: 604997
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOK2 PubMed Links

Gene Details

Gene
DOK2
Gene Name
docking protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317800.1 1120 Silent Mutation GCA,GCG A213A NP_001304729.1
NM_003974.3 1120 Silent Mutation GCA,GCG A367A NP_003965.2
NM_201349.2 1120 Silent Mutation GCA,GCG A213A NP_958728.1
XM_005273680.4 1120 Silent Mutation GCA,GCG A273A XP_005273737.1

View Full Product Details