Product Details

SNP ID
rs140268500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:132582900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTTGCTCGGGGACTGCCGTTCG[C/T]CCCTGGAGCTGAGTCCACGCTCAGA
Phenotype
MIM: 605211
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BARHL1 PubMed Links

Gene Details

Gene
BARHL1
Gene Name
BarH like homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020064.3 295 Missense Mutation CCC,TCC P35S NP_064448.1
Gene
CFAP77
Gene Name
cilia and flagella associated protein 77
There are no transcripts associated with this gene.

View Full Product Details