Product Details

SNP ID
rs140300211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97417016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGCATCTCAGTTTTGATTTTGA[A/G]TAGGGTGGCCAGAGAAGGCCTTACT
Phenotype
MIM: 611258
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TDRD7 PubMed Links

Gene Details

Gene
TDRD7
Gene Name
tudor domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302884.1 Intron NP_001289813.1
NM_014290.2 Intron NP_055105.2
XM_006717026.3 Intron XP_006717089.1

View Full Product Details