Product Details

SNP ID
rs142850734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35957274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGGGGATGAGTTTGTCTGAAAG[A/G]TCCTCTTTGTCTGCTCCCATGGAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2S2 PubMed Links

Gene Details

Gene
OR2S2
Gene Name
olfactory receptor family 2 subfamily S member 2 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019897.2 881 Silent Mutation GAC,GAT D275D NP_063950.2

View Full Product Details