Product Details

SNP ID
rs144789024
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:19116596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACAGGAGGGTGTGGCACGTGGT[C/T]TGGAGCTGCTGAGTCAGGTTGCGGG
Phenotype
MIM: 103195
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PLIN2 PubMed Links
Additional Information
For this assay, SNP(s) [rs2229536] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PLIN2
Gene Name
perilipin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122.3 988 Intron NP_001113.2
XM_017014259.1 988 Missense Mutation XP_016869748.1

View Full Product Details