Product Details

SNP ID
rs142520294
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136216736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATTTGCTGGCTGCCTGTGTTCA[C/T]AGCTCAGCCAACAATTGGTGTTGCC
Phenotype
MIM: 300163 MIM: 300930
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FHL1 PubMed Links

Gene Details

Gene
FHL1
Gene Name
four and a half LIM domains 1
There are no transcripts associated with this gene.

Gene
MAP7D3
Gene Name
MAP7 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173516.1 Intron NP_001166987.1
NM_001173517.1 Intron NP_001166988.1
NM_024597.3 Intron NP_078873.2
XM_005262472.1 Intron XP_005262529.1
XM_017029843.1 Intron XP_016885332.1

View Full Product Details