Product Details
- SNP ID
-
rs147816482
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:154507242 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTGTTCTGCACACCCTTGGCCCC[A/G]ACAAACACCCAGCTGTCCCGGAAGG
- Phenotype
-
MIM: 300492
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAM3A
PubMed Links
Gene Details
- Gene
- FAM3A
- Gene Name
- family with sequence similarity 3 member A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001171132.2 |
1273 |
Silent Mutation |
GTC,GTT |
V186V |
NP_001164603.1 |
NM_001171133.2 |
1273 |
Silent Mutation |
GTC,GTT |
V148V |
NP_001164604.1 |
NM_001171134.2 |
1273 |
Silent Mutation |
GTC,GTT |
V169V |
NP_001164605.1 |
NM_001282311.1 |
1273 |
Silent Mutation |
GTC,GTT |
V200V |
NP_001269240.1 |
NM_001282312.1 |
1273 |
Silent Mutation |
GTC,GTT |
V186V |
NP_001269241.1 |
NM_021806.3 |
1273 |
Silent Mutation |
GTC,GTT |
V186V |
NP_068578.2 |
XM_005274714.3 |
1273 |
Silent Mutation |
GTC,GTT |
V207V |
XP_005274771.1 |
XM_005274716.3 |
1273 |
Silent Mutation |
GTC,GTT |
V200V |
XP_005274773.1 |
XM_005277879.4 |
1273 |
Silent Mutation |
GTC,GTT |
V169V |
XP_005277936.1 |
XM_006724831.2 |
1273 |
Silent Mutation |
GTC,GTT |
V208V |
XP_006724894.1 |
XM_006724832.3 |
1273 |
Silent Mutation |
GTC,GTT |
V194V |
XP_006724895.1 |
XM_006724833.2 |
1273 |
Silent Mutation |
GTC,GTT |
V183V |
XP_006724896.1 |
XM_006724834.2 |
1273 |
Silent Mutation |
GTC,GTT |
V153V |
XP_006724897.1 |
XM_011531185.2 |
1273 |
Silent Mutation |
GTC,GTT |
V153V |
XP_011529487.1 |
XM_017029699.1 |
1273 |
Silent Mutation |
GTC,GTT |
V153V |
XP_016885188.1 |
XM_017029700.1 |
1273 |
Silent Mutation |
GTC,GTT |
V145V |
XP_016885189.1 |
XM_017029701.1 |
1273 |
Silent Mutation |
GTC,GTT |
V145V |
XP_016885190.1 |
XM_017029702.1 |
1273 |
Silent Mutation |
GTC,GTT |
V145V |
XP_016885191.1 |
View Full Product Details