Product Details

SNP ID
rs148556006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:20132201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTACCATGCTCTGGAAGCTCGCC[A/G]TATGCCTTCAGACTTCTAGCTTCGT
Phenotype
MIM: 300186
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF1AX PubMed Links

Gene Details

Gene
EIF1AX
Gene Name
eukaryotic translation initiation factor 1A, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001412.3 Intron NP_001403.1
Gene
EIF1AX-AS1
Gene Name
EIF1AX antisense RNA 1
There are no transcripts associated with this gene.

Gene
SCARNA9L
Gene Name
small Cajal body-specific RNA 9-like
There are no transcripts associated with this gene.

View Full Product Details