Product Details

SNP ID
rs150263726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106942929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAATTCTTCAGGAAGCAGGGACCC[C/T]TGGCGTTGGTCTTTGGCCCCTGGAT
Phenotype
MIM: 300970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MORC4 PubMed Links

Gene Details

Gene
MORC4
Gene Name
MORC family CW-type zinc finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085354.2 2237 Silent Mutation CAA,CAG Q654Q NP_001078823.1
NM_024657.4 2237 Silent Mutation CAA,CAG Q654Q NP_078933.3
XM_005262190.3 2237 Silent Mutation CAA,CAG Q562Q XP_005262247.1
XM_006724691.2 2237 Silent Mutation CAA,CAG Q532Q XP_006724754.1
XM_011531027.2 2237 Silent Mutation CAA,CAG Q477Q XP_011529329.1
XM_017029844.1 2237 Silent Mutation CAA,CAG Q562Q XP_016885333.1

View Full Product Details