Product Details

SNP ID
rs150736453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106942681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGCTGCAGCAGGGATTGCAGCA[A/G]AGGCCACAGAATAAGGCACTGGGTT
Phenotype
MIM: 300970
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MORC4 PubMed Links

Gene Details

Gene
MORC4
Gene Name
MORC family CW-type zinc finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085354.2 2485 Missense Mutation TCT,TTT S737F NP_001078823.1
NM_024657.4 2485 Missense Mutation TCT,TTT S737F NP_078933.3
XM_005262190.3 2485 Missense Mutation TCT,TTT S645F XP_005262247.1
XM_006724691.2 2485 Missense Mutation TCT,TTT S615F XP_006724754.1
XM_011531027.2 2485 Missense Mutation TCT,TTT S560F XP_011529329.1
XM_017029844.1 2485 Missense Mutation TCT,TTT S645F XP_016885333.1

View Full Product Details