Product Details

SNP ID
rs11575848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31719166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAAGGCCCAGGGCTGGAGTGGGC[T/C]GGGGTCCTGCGCTGTTGCAGTTGTC
Phenotype
MIM: 606520 MIM: 604744 MIM: 610435 MIM: 606038 MIM: 610437 MIM: 611404
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
C6orf25 PubMed Links

Gene Details

Gene
C6orf25
Gene Name
chromosome 6 open reading frame 25
There are no transcripts associated with this gene.

Gene
DDAH2
Gene Name
dimethylarginine dimethylaminohydrolase 2
There are no transcripts associated with this gene.

Gene
LY6G6C
Gene Name
lymphocyte antigen 6 complex, locus G6C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025261.2 363 Missense Mutation CAG,CGG Q103R NP_079537.1
Gene
LY6G6D
Gene Name
lymphocyte antigen 6 complex, locus G6D
There are no transcripts associated with this gene.

Gene
LY6G6E
Gene Name
lymphocyte antigen 6 complex, locus G6E (pseudogene)
There are no transcripts associated with this gene.

Gene
LY6G6F
Gene Name
lymphocyte antigen 6 complex, locus G6F
There are no transcripts associated with this gene.

View Full Product Details