Product Details

SNP ID
rs7292477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24938217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCACAGCCCTTTGGGGGCCACA[G/T]CCCAAGACAGGAGGAAAATTGCATT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM211 PubMed Links

Gene Details

Gene
TMEM211
Gene Name
transmembrane protein 211
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001663.1 84 Missense Mutation GAT,GCT D20A NP_001001663.1
XM_011530094.2 84 Missense Mutation GAT,GCT D81A XP_011528396.1

View Full Product Details