Product Details
- SNP ID
-
rs11223758
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:134336099 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGTGTTTTTAGAGGCTCCTGCTAA[C/T]GAGGTAGATATTTCTTGACTCGTGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GLB1L2
PubMed Links
Gene Details
- Gene
- GLB1L2
- Gene Name
- galactosidase beta 1 like 2
View Full Product Details