Product Details

SNP ID
rs181154367
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:66814175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATCCAAGCAAAGTATCCATTAT[A/G]TCTCCCTGAAAAAAAAAAGTCACAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR78 PubMed Links

Gene Details

Gene
WDR78
Gene Name
WD repeat domain 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024763.4 2316 Silent Mutation GAC,GAT D834D NP_079039.4
NM_207014.2 2316 Intron NP_996897.2
XM_005271212.3 2316 Silent Mutation GAC,GAT D769D XP_005271269.1
XM_011542161.2 2316 Silent Mutation GAC,GAT D805D XP_011540463.1
XM_011542162.2 2316 Silent Mutation GAC,GAT D801D XP_011540464.1
XM_017002353.1 2316 Silent Mutation GAC,GAT D736D XP_016857842.1
XM_017002354.1 2316 Silent Mutation GAC,GAT D707D XP_016857843.1
XM_017002355.1 2316 Silent Mutation GAC,GAT D580D XP_016857844.1
XM_017002356.1 2316 Silent Mutation GAC,GAT D545D XP_016857845.1
XM_017002357.1 2316 Intron XP_016857846.1

View Full Product Details