Product Details

SNP ID
rs182453905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:6849853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTGGCACCTGGCCAGAGTAACTA[C/T]GGTTCACTAAAACTATTCTGGATTA
Phenotype
MIM: 611787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CMPK2 PubMed Links

Gene Details

Gene
CMPK2
Gene Name
cytidine/uridine monophosphate kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256477.1 1470 Intron NP_001243406.1
NM_001256478.1 1470 Intron NP_001243407.1
NM_207315.3 1470 Silent Mutation NP_997198.2
Gene
NRIR
Gene Name
negative regulator of interferon response (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details