Product Details
- SNP ID
-
rs182611179
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:9772807 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCCCAGTTTCCTTGCTCTGGATA[C/T]ACGTGGTACGACCGGATGAAAGTCG
- Phenotype
-
MIM: 616161
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DHRS7C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2280490] are located under a probe and SNP(s) [rs2280489] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DHRS7C
- Gene Name
- dehydrogenase/reductase 7C
View Full Product Details