Product Details

SNP ID
rs185607856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67065920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTACACCCCCACGGTGTTTGAG[C/T]GGTACATGGTCAACCTGCAAGTGAA
Phenotype
MIM: 605781
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RHOD PubMed Links

Gene Details

Gene
RHOD
Gene Name
ras homolog family member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300886.1 242 Intron NP_001287815.1
NM_014578.3 242 Missense Mutation CGG,TGG R53W NP_055393.1

View Full Product Details