Product Details
- SNP ID
-
rs186130633
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:65524242 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGAGAACGGCTTTGGCACCAGGA[C/T]GTCATTCAAGTCCACGTTTTCTGGT
- Phenotype
-
MIM: 300964
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LAS1L
PubMed Links
Gene Details
- Gene
- LAS1L
- Gene Name
- LAS1 like, ribosome biogenesis factor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001170649.1 |
1139 |
Missense Mutation |
ATC,GTC |
I355V |
NP_001164120.1 |
NM_001170650.1 |
1139 |
Missense Mutation |
ATC,GTC |
I313V |
NP_001164121.1 |
NM_031206.4 |
1139 |
Missense Mutation |
ATC,GTC |
I372V |
NP_112483.1 |
XM_005262301.1 |
1139 |
Missense Mutation |
ATC,GTC |
I372V |
XP_005262358.1 |
XM_005262304.1 |
1139 |
Missense Mutation |
ATC,GTC |
I372V |
XP_005262361.1 |
XM_005262305.3 |
1139 |
Missense Mutation |
ATC,GTC |
I372V |
XP_005262362.1 |
XM_005262306.3 |
1139 |
Missense Mutation |
ATC,GTC |
I372V |
XP_005262363.1 |
XM_011531045.1 |
1139 |
Missense Mutation |
ATC,GTC |
I330V |
XP_011529347.1 |
XM_011531046.1 |
1139 |
Missense Mutation |
ATC,GTC |
I372V |
XP_011529348.1 |
XM_017029877.1 |
1139 |
Missense Mutation |
ATC,GTC |
I355V |
XP_016885366.1 |
XM_017029878.1 |
1139 |
Missense Mutation |
ATC,GTC |
I355V |
XP_016885367.1 |
XM_017029879.1 |
1139 |
Missense Mutation |
ATC,GTC |
I70V |
XP_016885368.1 |
XM_017029880.1 |
1139 |
Missense Mutation |
ATC,GTC |
I53V |
XP_016885369.1 |
View Full Product Details