Product Details
- SNP ID
-
rs185796187
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:20785330 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCGAGTGCGCGGAGGAGCCGCCGC[A/C]GCCGCCCCCGCCGCCGCCGCCGCTG
- Phenotype
-
MIM: 608306
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SP8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs372591893] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SP8
- Gene Name
- Sp8 transcription factor
View Full Product Details