Product Details

SNP ID
rs186591921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48626290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAGCATCATGGACAGAGGCAAA[G/T]AGATGCTTCTTGGTGATGGATGCAT
Phenotype
MIM: 610068
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR6824 PubMed Links

Gene Details

Gene
MIR6824
Gene Name
microRNA 6824
There are no transcripts associated with this gene.

Gene
SLC26A6
Gene Name
solute carrier family 26 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040454.1 2293 Silent Mutation CTA,CTC L710L NP_001035544.1
NM_001281732.1 2293 Silent Mutation CTA,CTC L695L NP_001268661.1
NM_001281733.1 2293 Silent Mutation CTA,CTC L623L NP_001268662.1
NM_022911.2 2293 Silent Mutation CTA,CTC L731L NP_075062.2
NM_134263.2 2293 Silent Mutation CTA,CTC L730L NP_599025.2
NM_134426.2 2293 Silent Mutation CTA,CTC L712L NP_602298.2
Gene
TMEM89
Gene Name
transmembrane protein 89
There are no transcripts associated with this gene.

View Full Product Details