Product Details

SNP ID
rs187705818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32360932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAGGTGTTCCTCCTCTGATGCC[A/G]GGAATGCCACCAGTTATGCCAGGCA
Phenotype
MIM: 603428
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF207 PubMed Links

Gene Details

Gene
ZNF207
Gene Name
zinc finger protein 207
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032293.2 796 Intron NP_001027464.1
NM_001098507.1 796 Intron NP_001091977.1
NM_003457.3 796 Intron NP_003448.1
XM_011525185.2 796 Silent Mutation CCA,CCG P118P XP_011523487.1
XM_017025019.1 796 Silent Mutation CCA,CCG P172P XP_016880508.1
XM_017025020.1 796 Intron XP_016880509.1

View Full Product Details