Product Details

SNP ID
rs188735942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1556772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCTTGCTGCGCGTGGCGCGGAT[G/T]ATGGAGAAGTGTTCGGCCGCCGACA
Phenotype
MIM: 611009
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MEX3D PubMed Links

Gene Details

Gene
MEX3D
Gene Name
mex-3 RNA binding family member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174118.1 747 Silent Mutation NP_001167589.1
NM_203304.3 747 Silent Mutation NP_976049.3
XM_017026811.1 747 Silent Mutation XP_016882300.1

View Full Product Details