Product Details

SNP ID
rs188687743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99547487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCATATACTCTGCTCAATACTTT[C/T]CCTTGAGAAAGCCAACGAACTTCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM200A PubMed Links

Gene Details

Gene
FAM200A
Gene Name
family with sequence similarity 200 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145111.3 1105 Silent Mutation GGA,GGG G307G NP_659802.1
XM_011515911.2 1105 Silent Mutation GGA,GGG G307G XP_011514213.1
XM_011515913.2 1105 Silent Mutation GGA,GGG G307G XP_011514215.1

View Full Product Details