Product Details

SNP ID
rs188937673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:56441605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCCCTGTTTCCAGAAGCTCTTT[G/T]AGCAGGTCTGAGGAATTGGCCAGCG
Phenotype
MIM: 131330
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929415 PubMed Links

Gene Details

Gene
LOC101929415
Gene Name
uncharacterized LOC101929415
There are no transcripts associated with this gene.

Gene
PENK
Gene Name
proenkephalin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135690.2 696 Silent Mutation CTA,CTC L157L NP_001129162.1

View Full Product Details