Product Details

SNP ID
rs190435213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81525051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGACAGCTTGAGAAAGCCTTAAG[A/T]GTTTGAGACTGAGTTTGATGATACT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FABP12 PubMed Links

Gene Details

Gene
FABP12
Gene Name
fatty acid binding protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105281.1 418 Missense Mutation ACT,TCT T140S NP_001098751.1
XM_005251285.4 418 Missense Mutation ACT,TCT T140S XP_005251342.1
XM_006716465.3 418 Missense Mutation ACT,TCT T140S XP_006716528.1
XM_011517577.2 418 Missense Mutation ACT,TCT T140S XP_011515879.1

View Full Product Details