Product Details

SNP ID
rs191617125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45795924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCGCCCTCCTCCTCCTCCTCT[C/G]TGGCTCCCAGGGCTTGTTCCTGGGC
Phenotype
MIM: 609857 MIM: 607548
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DMWD PubMed Links

Gene Details

Gene
DMWD
Gene Name
dystrophia myotonica, WD repeat containing
There are no transcripts associated with this gene.

Gene
RSPH6A
Gene Name
radial spoke head 6 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030785.3 2270 Missense Mutation ACA,AGA T700R NP_110412.1
XM_011527351.1 2270 Intron XP_011525653.1

View Full Product Details