Product Details

SNP ID
rs191495317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:67066172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGCCATGTCGTCCATCCTGCC[C/T]TTCACTCCCCCGATCGTGAAGCGCC
Phenotype
MIM: 603109
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102723493 PubMed Links

Gene Details

Gene
LOC102723493
Gene Name
uncharacterized LOC102723493
There are no transcripts associated with this gene.

Gene
SMAD3
Gene Name
SMAD family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145102.1 316 Intron NP_001138574.1
NM_001145103.1 316 Intron NP_001138575.1
NM_001145104.1 316 Intron NP_001138576.1
NM_005902.3 316 Silent Mutation CCC,CCT P6P NP_005893.1
XM_011521559.2 316 Silent Mutation CCC,CCT P6P XP_011519861.1
XM_011521560.1 316 Intron XP_011519862.1

View Full Product Details