Product Details

SNP ID
rs191231311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101087324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTGCCTGCAGCATGGTTTCCCA[C/T]GGGTCCTCGCCCTCCCTCCTGGAGG
Phenotype
MIM: 616996
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM56 PubMed Links

Gene Details

Gene
TRIM56
Gene Name
tripartite motif containing 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030961.2 309 Silent Mutation CAC,CAT H4H NP_112223.1
XM_011516589.2 309 Silent Mutation CAC,CAT H4H XP_011514891.1

View Full Product Details