Product Details

SNP ID
rs191789132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119085422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTAGTCTCAGCATCGGCTCCAGC[A/G]TTGCTCTTAGTTTTCAGCTCTTTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1210 PubMed Links

Gene Details

Gene
KIAA1210
Gene Name
KIAA1210
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020721.1 4398 Silent Mutation AAC,AAT N1603N NP_065772.1
XM_006724774.3 4398 Silent Mutation AAC,AAT N1427N XP_006724837.1
XM_017029688.1 4398 Silent Mutation AAC,AAT N1442N XP_016885177.1
XM_017029689.1 4398 Silent Mutation AAC,AAT N1376N XP_016885178.1

View Full Product Details