Product Details

SNP ID
rs192275688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92468856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTTGATGTCATTAACATCAGAT[C/T]GTGGGCTAGCAGGCCGGTCCAGGGG
Phenotype
MIM: 604659 MIM: 614518
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERVW-1 PubMed Links

Gene Details

Gene
ERVW-1
Gene Name
endogenous retrovirus group W member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130925.1 Intron NP_001124397.1
NM_014590.3 Intron NP_055405.3
Gene
GATAD1
Gene Name
GATA zinc finger domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021167.4 Intron NP_066990.3

View Full Product Details