Product Details

SNP ID
rs192536705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143222342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTCTGCTATTCCGAATTGTTTA[C/T]AACCAAAACTCAGTGTATATCCTCT
Phenotype
MIM: 613964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAS2R40 PubMed Links

Gene Details

Gene
TAS2R40
Gene Name
taste 2 receptor member 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176882.1 264 Silent Mutation TAC,TAT Y88Y NP_795363.1

View Full Product Details