Product Details

SNP ID
rs6722317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:168790974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTTGAGAGTATTTTTAATCAAT[A/G]TATTTGTTTTCTTTTTTTAAATTTT
Phenotype
MIM: 607496
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CERS6-AS1 PubMed Links

Gene Details

Gene
CERS6-AS1
Gene Name
CERS6 antisense RNA 1
There are no transcripts associated with this gene.

Gene
NOSTRIN
Gene Name
nitric oxide synthase trafficking
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039724.3 Intron NP_001034813.2
NM_001171631.1 Intron NP_001165102.1
NM_001171632.1 Intron NP_001165103.1
NM_052946.3 Intron NP_443178.2
XM_005246270.1 Intron XP_005246327.1
XM_006712233.3 Intron XP_006712296.1
XM_011510540.2 Intron XP_011508842.1
XM_011510543.2 Intron XP_011508845.1
XM_017003278.1 Intron XP_016858767.1
XM_017003279.1 Intron XP_016858768.1
XM_017003280.1 Intron XP_016858769.1

View Full Product Details