Product Details

SNP ID
rs7200729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87949449 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACTGCCGGAGGGAATATTCTGTG[G/T]CAGCTATCGCACGCACCCTTTCCCT
Phenotype
MIM: 611564
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BANP PubMed Links

Gene Details

Gene
BANP
Gene Name
BTG3 associated nuclear protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173539.1 234 Intron NP_001167010.1
NM_001173540.1 234 Intron NP_001167011.1
NM_001173541.1 234 Intron NP_001167012.1
NM_001173542.1 234 Intron NP_001167013.1
NM_001173543.1 234 Intron NP_001167014.1
NM_017869.3 234 Intron NP_060339.2
NM_079837.2 234 Intron NP_524576.2
XM_005256008.3 234 Intron XP_005256065.1
XM_005256009.3 234 Intron XP_005256066.1
XM_011523170.2 234 Intron XP_011521472.1
XM_011523172.2 234 Intron XP_011521474.1
XM_011523174.2 234 Intron XP_011521476.1
XM_011523175.2 234 Intron XP_011521477.1
XM_011523176.2 234 UTR 5 XP_011521478.1
XM_011523177.2 234 Intron XP_011521479.1
XM_011523178.2 234 Intron XP_011521480.1
XM_011523179.2 234 Intron XP_011521481.1
XM_011523180.2 234 Intron XP_011521482.1
XM_011523181.2 234 Intron XP_011521483.1
XM_011523182.1 234 Intron XP_011521484.1
XM_011523183.2 234 Intron XP_011521485.1
XM_011523184.2 234 Intron XP_011521486.1
XM_011523186.2 234 Intron XP_011521488.1
XM_017023378.1 234 Intron XP_016878867.1
XM_017023379.1 234 Intron XP_016878868.1
XM_017023380.1 234 Intron XP_016878869.1
XM_017023381.1 234 Intron XP_016878870.1
XM_017023382.1 234 Intron XP_016878871.1
XM_017023383.1 234 Intron XP_016878872.1

View Full Product Details