Product Details

SNP ID
rs738145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38782388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTACGATCATGTTCTTAAAACA[C/T]GTATTTCATGTGGGCCAGGCCTTCG
Phenotype
MIM: 610565
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAL4 PubMed Links

Gene Details

Gene
DNAL4
Gene Name
dynein axonemal light chain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005740.2 Intron NP_005731.1

View Full Product Details