Product Details
- SNP ID
-
rs7931491
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:9281574 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTACTTTTCAACTTTGACAACTAAT[A/G]ACAGTCATGGGTGAAGGTAAAACTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM41B
PubMed Links
Gene Details
- Gene
- TMEM41B
- Gene Name
- transmembrane protein 41B
View Full Product Details