Product Details

SNP ID
rs201474003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:27116268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATAAGCTCCTCTGCCACTCTTCC[C/T]CCCATACTAACATCCATTTGTGCAA
Phenotype
MIM: 607472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
YME1L1 PubMed Links

Gene Details

Gene
YME1L1
Gene Name
YME1 like 1 ATPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253866.1 2461 Silent Mutation GGA,GGG G566G NP_001240795.1
NM_014263.3 2461 Silent Mutation GGA,GGG G599G NP_055078.1
NM_139312.2 2461 Silent Mutation GGA,GGG G656G NP_647473.1
XM_011519300.2 2461 Silent Mutation GGA,GGG G623G XP_011517602.1

View Full Product Details