Product Details
- SNP ID
-
rs200152066
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:63558453 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACGAAGTGCTCGCAGTTGTCACTG[G/T]TCAGCGAATAAGGCAACTCCTGCCC
- Phenotype
-
MIM: 613866
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HRASLS2
PubMed Links
Gene Details
- Gene
- HRASLS2
- Gene Name
- HRAS like suppressor 2
There are no transcripts associated with this gene.
View Full Product Details