Product Details

SNP ID
rs200297051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62835460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTACCCTGTGCAGGTGACAAGCT[C/T]CTGTGATCCTGGGAGAAACTGTATG
Phenotype
MIM: 603189
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105369332 PubMed Links

Gene Details

Gene
LOC105369332
Gene Name
uncharacterized LOC105369332
There are no transcripts associated with this gene.

Gene
RNU2-2P
Gene Name
RNA, U2 small nuclear 2, pseudogene
There are no transcripts associated with this gene.

Gene
STX5
Gene Name
syntaxin 5
There are no transcripts associated with this gene.

Gene
WDR74
Gene Name
WD repeat domain 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307977.1 678 Missense Mutation AAG,GAG K197E NP_001294906.1
NM_018093.3 678 Missense Mutation AAG,GAG K197E NP_060563.2
XM_005274055.2 678 Missense Mutation AAG,GAG K197E XP_005274112.1

View Full Product Details