Product Details

SNP ID
rs199988593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18534378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTTTCTTCTTATTGTTTACAAT[A/G]CTGTCAACCATGTCAGCTACTGATA
Phenotype
MIM: 601387 MIM: 610985
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSG101 PubMed Links

Gene Details

Gene
TSG101
Gene Name
tumor susceptibility 101
There are no transcripts associated with this gene.

Gene
UEVLD
Gene Name
UEV and lactate/malate dehyrogenase domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040697.2 3123 Silent Mutation AGC,AGT S400S NP_001035787.1
NM_001261382.1 3123 Silent Mutation AGC,AGT S378S NP_001248311.1
NM_001261383.1 3123 Intron NP_001248312.1
NM_001261384.1 3123 Silent Mutation AGC,AGT S270S NP_001248313.1
NM_001261385.1 3123 Intron NP_001248314.1
NM_001261386.1 3123 Intron NP_001248315.1
NM_001297771.1 3123 Intron NP_001284700.1
NM_018314.4 3123 Intron NP_060784.3
XM_006718255.2 3123 Silent Mutation AGC,AGT S177S XP_006718318.1
XM_011520201.2 3123 Silent Mutation AGC,AGT S177S XP_011518503.1
XM_017017985.1 3123 Silent Mutation AGC,AGT S270S XP_016873474.1
XM_017017986.1 3123 Intron XP_016873475.1

View Full Product Details