Product Details

SNP ID
rs200172846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4803439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATACACTGACCCTTTGCTCAGGG[C/G]ATGTTTCCACAACATCCTTGGATAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR52R1 PubMed Links

Gene Details

Gene
OR52R1
Gene Name
olfactory receptor family 52 subfamily R member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005177.3 942 Silent Mutation ATC,ATG I314M NP_001005177.3

View Full Product Details