Product Details

SNP ID
rs199730490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:95067689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGATCTCGCTATAGGGGTTCTCGC[C/T]ATAGCCGGTCTCCCTACAGCCGATC
Phenotype
MIM: 603269
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRSF8 PubMed Links

Gene Details

Gene
SRSF8
Gene Name
serine and arginine rich splicing factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032102.3 813 Missense Mutation CAT,TAT H155Y NP_115285.1
XM_017017142.1 813 Missense Mutation CAT,TAT H155Y XP_016872631.1

View Full Product Details