Product Details

SNP ID
rs201997939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119025739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCGCGAGAGGGTGAAGAGCCA[C/G]AGTCCAGAAAGCAACATCCTAGAGG
Phenotype
MIM: 180465 MIM: 602671 MIM: 610971
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC84 PubMed Links

Gene Details

Gene
CCDC84
Gene Name
coiled-coil domain containing 84
There are no transcripts associated with this gene.

Gene
MIR3656
Gene Name
microRNA 3656
There are no transcripts associated with this gene.

Gene
RPS25
Gene Name
ribosomal protein S25
There are no transcripts associated with this gene.

Gene
SLC37A4
Gene Name
solute carrier family 37 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164277.1 1444 Intron NP_001157749.1
NM_001164278.1 1444 Missense Mutation CTG,GTG L335V NP_001157750.1
NM_001164279.1 1444 Intron NP_001157751.1
NM_001164280.1 1444 Intron NP_001157752.1
NM_001467.5 1444 Intron NP_001458.1
Gene
TRAPPC4
Gene Name
trafficking protein particle complex 4
There are no transcripts associated with this gene.

View Full Product Details