Product Details

SNP ID
rs202023755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73951088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGCCATGGGCCAGGATTATTAC[A/T]CTGTGCTCGGGATCACTCGCAATTC
Phenotype
MIM: 610263
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJB13 PubMed Links

Gene Details

Gene
DNAJB13
Gene Name
DnaJ heat shock protein family (Hsp40) member B13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153614.3 468 Missense Mutation ACT,TCT T7S NP_705842.2
XM_005273984.3 468 Missense Mutation ACT,TCT T7S XP_005274041.1
XM_011545004.2 468 UTR 5 XP_011543306.1
XM_011545005.2 468 UTR 5 XP_011543307.1
XM_011545007.2 468 Intron XP_011543309.1
XM_011545009.2 468 UTR 5 XP_011543311.1
XM_011545013.2 468 UTR 5 XP_011543315.1
XM_011545014.2 468 UTR 5 XP_011543316.1
XM_011545015.2 468 Intron XP_011543317.1
XM_017017675.1 468 Intron XP_016873164.1
XM_017017676.1 468 Intron XP_016873165.1
XM_017017677.1 468 Intron XP_016873166.1
XM_017017678.1 468 Intron XP_016873167.1
XM_017017679.1 468 UTR 5 XP_016873168.1

View Full Product Details