Product Details
- SNP ID
-
rs202076454
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:72577554 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTCACGGTTGGAGGCCACGCGCT[C/T]GTACAGCTCTGCCGCTTTGGGGAAC
- Phenotype
-
MIM: 602658
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC01537
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1980091] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC01537
- Gene Name
- long intergenic non-protein coding RNA 1537
There are no transcripts associated with this gene.
- Gene
- PDE2A
- Gene Name
- phosphodiesterase 2A
View Full Product Details